2-sec-Butyl-3-methoxypyrazine, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-(sec-Butyl)-3-methoxypyrazine

M-Clarity™ quality level = MQ300

W343307-100G-K W343307-1KG-K
SIALW343307-100G-KEA 647 EUR
SIALW343307-100G-K SIALW343307-1KG-K
2-sec-Butyl-3-methoxypyrazine, Sigma-Aldrich®
2-sec-Butyl-3-methoxypyrazine
Formula: C₉H₁₄N₂O
MW: 166.2203 g/mol
MDL Number: MFCD00006136
CAS Number: 24168-70-5
EINECS: 246-050-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR