Diethyl difluoromalonate ≥97%
Supplier: Thermo Fisher Scientific
Warning
L16042.03 L16042.06
L16042.03EA 80.5 EUR
L16042.03 L16042.06
Diethyl difluoromalonate ≥97%
Diethyl difluoromalonate
Purity: 97% CAS No.: 680-65-9 Molecular Formula: C7H10F2O4 Formula Weight: 196.15 Refractive Index: 1.3800 Boiling Point: 94-95°/23mm MDL No.: MFCD00221617
Formula: C₇H₁₀F₂O₄
MW: 196.15 g/mol
Boiling Pt: 94…95 °C (23 mmHg)
Density: 1.179
Storage Temperature: Ambient
MDL Number: MFCD00221617
CAS Number: 680-65-9