5-Methylpyrazine-2-carboxylic acid

Supplier: Thermo Scientific

SB01107.1 SB01107EA SB01107.50
MAYBSB01107.1EA 0 EUR
MAYBSB01107.1 MAYBSB01107EA MAYBSB01107.50
5-Methylpyrazine-2-carboxylic acid
5-Methylpyrazine-2-carboxylic acid
Formula: C₆H₆N₂O₂
MW: 138.13 g/mol
MDL Number: MFCD00068241
CAS Number: 5521-55-1
EINECS: 413-260-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR