(±)-3-Hydroxytetrahydrofuran 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
121940050
121940250
121941000
ACRO121940050EA
36.4
EUR
ACRO121940050
ACRO121940250
ACRO121941000
(±)-3-Hydroxytetrahydrofuran 97%
(±)-3-Hydroxytetrahydrofuran
Formula:
C₄H₈O₂ Boiling Pt: 181 °C (760 mmHg) Density: 1.09 g/cm³ Flash Pt: 81 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005374 CAS Number: 453-20-3 EINECS: 207-219-2 |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4490 to 1.4510 (20°C, 589 nm) |
GC | ≥96.0 % |
UV | at 240 nm A: ≤0.05 |
Titration after Ion exchange | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...