4-(2-Pyridylazo)resorcinol monosodim salt monohydrate
Supplier: Thermo Fisher Scientific
Warning
Synonyms: PAR monosodium salt monohydrate, 4-(2-Pyridylazo)resorcinol monosodium salt monohydrate, PAR
A18103.03 A18103.06 A18103.14
A18103.03EA 27.8 EUR
A18103.03 A18103.06 A18103.14
4-(2-Pyridylazo)resorcinol monosodim salt monohydrate
4-(2-Pyridylazo)resorcinol monosodim salt monohydrate
CAS No.: 16593-81-0 Molecular Formula: C11H8N3NaO2.H2O Formula Weight: 255.21 (237.19anhy) Melting Point: 300° Beilstein Reference: 3825637 MDL No.: MFCD00150712
Formula: C₁₁H₈N₃NaO₂·1H₂O
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00150712
CAS Number: 16593-81-0
EINECS: 236-339-8