2-Hydroxycyclohexanone dimer ≥90%
Supplier: Thermo Fisher Scientific
|
Warning
|
H27199.14
H27199.06
H27199.14EA
195
EUR
H27199.14
H27199.06
2-Hydroxycyclohexanone dimer ≥90%
2-Hydroxycyclohexanone dimer
Purity: 90+%
CAS No.: 30282-14-5
Molecular Formula: C12H20O4
Formula Weight: 228.28
Flash Point: 79°(174°F)
Melting Point: 100-107°
Boiling Point: 83-86°/13mm
UN No.: 1325
MDL No.: MFCD00151157
Formula:
C₆H₁₀O₂ Boiling Pt: 83…86 °C (13 mmHg) Melting Pt: 100…107 °C Flash Pt: 79 °C (174 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00151157 CAS Number: 30282-14-5 UN: 1325 ADR: 4.1,III |