Tetrabutylammonium acetate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Tetra-n-butylammonium acetate

86849-10G 86849-50G
SIAL86849-10GEA 151 EUR
SIAL86849-10G SIAL86849-50G
Tetrabutylammonium acetate, Sigma-Aldrich®
Tetrabutylammonium acetate
Formula: C₁₈H₃₉NO₂
MDL Number: MFCD00043208
CAS Number: 10534-59-5
EINECS: 234-101-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR