N-Methyl-1,3-propanediamine 99%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
N-Methylpropane-1,3-diamine, 3-Methylamino propylamine
B23319.22
B23319.36
B23319.22EA
45.4
EUR
B23319.22
B23319.36
N-Methyl-1,3-propanediamine 99%
N-Methyl-1,3-propanediamine
Purity: 99%
CAS No.: 6291-84-5
Molecular Formula: CH3NH(CH2)3NH2
Formula Weight: 88.15
Flash Point: 35°(96°F)
Refractive Index: 1.4475
Melting Point: -72°
Boiling Point: 139-141°
Beilstein Reference: 878143
UN No.: 2734
MDL No.: MFCD00008209
Formula:
C₄H₁₂N₂ MW: 88.15 g/mol Boiling Pt: 139…141 °C Melting Pt: –72 °C Density: 0.844 Flash Pt: 35 °C (96 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008209 CAS Number: 6291-84-5 EINECS: 228-544-6 ADR: 8 II |