4-Amino-6-chlorobenzene-1,3-disulphonamide, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

308560-5G 308560-100G
SIAL308560-5GEA 26.5 EUR
SIAL308560-5G SIAL308560-100G
4-Amino-6-chlorobenzene-1,3-disulphonamide, Sigma-Aldrich®
4-Amino-6-chlorobenzene-1,3-disulphonamide
Formula: C₆H₈ClN₃O₄S₂
MDL Number: MFCD00007933
CAS Number: 121-30-2
EINECS: 204-463-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR