4,4'-(Hexafluoroisopropylidene)diphenol ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
4,4′-(Hexafluoroisopropylidene)diphenol, 2,2-Bis(4-hydroxyphenyl)hexafluoropropane, 4,4'-(Perfluoropropane-2,2-diyl)diphenol
A18370.06
A18370.14
A18370.22
A18370.06EA
36.4
EUR
A18370.06
A18370.14
A18370.22
4,4'-(Hexafluoroisopropylidene)diphenol ≥98%
4,4'-(Hexafluoroisopropylidene)diphenol
Purity: 98%
CAS No.: 1478-61-1
Molecular Formula: C15H10F6O2
Formula Weight: 336.24
Melting Point: 158-162°
Boiling Point: 400°
MDL No.: MFCD00000439
Formula:
C₁₅H₁₀F₆O₂ Boiling Pt: 400 °C Melting Pt: 158…162 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000439 CAS Number: 1478-61-1 EINECS: 216-036-7 |