O-Benzylhydroxylamine hydrochloride

Supplier: Thermo Scientific

TL00614.1 TL00614.10 TL00614.50
MAYBTL00614.1EA 0 EUR
MAYBTL00614.1 MAYBTL00614.10 MAYBTL00614.50
O-Benzylhydroxylamine hydrochloride
O-Benzylhydroxylamine hydrochloride
Formula: C₇H₁₀ClNO
MW: 159.62 g/mol
MDL Number: MFCD00012952
CAS Number: 2687-43-6
EINECS: 220-249-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR