Hydroxylamine-O-sulphonic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms: Amino hydrogen sulphate
M-Clarity™ quality level = MQ200
480975-25G 480975-5G
SIAL480975-25GEA 133 EUR
SIAL480975-25G SIAL480975-5G
Hydroxylamine-O-sulphonic acid, Sigma-Aldrich®
Hydroxylamine-O-sulphonic acid
Formula: H₃NO₄S
MDL Number: MFCD00011604
CAS Number: 2950-43-8
EINECS: 220-971-6
UN: 3260
ADR: 8,III