4'-Aminobenzo-18-crown-6 (4'-aminobenzo-1,4,7,10,13,16-hexoxacyclooctadecane), Sigma-Aldrich®
Supplier: Merck
Synonyms: 2,3,5,6,8,9,11,12,14,15-Decahydrobenzo[b][1,4,7,10,13,16]hexaoxacyclooctadecin-18-amine
M-Clarity™ quality level = MQ100
06913-100MG 06913-500MG
SIAL06913-100MGEA 76.1 EUR
SIAL06913-100MG SIAL06913-500MG
4'-Aminobenzo-18-crown-6 (4'-aminobenzo-1,4,7,10,13,16-hexoxacyclooctadecane), Sigma-Aldrich®
4'-Aminobenzo-18-crown-6 (4'-Aminobenzo-1,4,7,10,13,16-hexoxacyclooctadecane)
Formula: C₁₆H₂₅NO₆
MDL Number: MFCD00210009
CAS Number: 68941-06-0