Sulphur trioxide pyridine complex, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
S7556-100G S7556-25G
SIALS7556-100GEA 105 EUR
SIALS7556-100G SIALS7556-25G
Sulphur trioxide pyridine complex, Sigma-Aldrich®
Sulphur trioxide pyridine complex
Formula: C₅H₅NO₃S
MW: 159.16 g/mol
MDL Number: MFCD00012437
CAS Number: 26412-87-3
EINECS: 247-683-3
UN: 3261
ADR: 8 II