cis-2,6-Dimethylmorpholine ≥97%
Supplier: Thermo Fisher Scientific
|
Danger
|
L13007.09
L13007.18
L13007.09EA
44.1
EUR
L13007.09
L13007.18
cis-2,6-Dimethylmorpholine ≥97%
cis-2,6-Dimethylmorpholine
Purity: 97%
CAS No.: 6485-55-8
Molecular Formula: C6H13NO
Formula Weight: 115.18
Flash Point: 48°(118°F)
Refractive Index: 1.4460
Melting Point: -85°
Boiling Point: 146-147°
Beilstein Reference: 878182
UN No.: 1992
MDL No.: MFCD00078428
Formula:
C₆H₁₃NO Boiling Pt: 146…147 °C Melting Pt: –85 °C Density: 0.935 Flash Pt: 48 °C (118 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00078428 CAS Number: 6485-55-8 EINECS: 229-353-0 UN: 1992 ADR: 3,III |