2-Vinylpyrazine ≥98% stabilised
Supplier: Thermo Fisher Scientific
|
Warning
|
Stabilization:
Stabilised with hydroquinone (HQ)
Stabilized with ca 0.1% hydroquinone
Purity: 98%
CAS No.: 4177-16-6
Molecular Formula: C6H6N2
Formula Weight: 106.13
Storage Temperature: -30°C to -10°C
Flash Point: 60°(140°F)
Refractive Index: 1.5560
Boiling Point: 163-164°
Beilstein Reference: 105566
UN No.: 1993
MDL No.: MFCD00051591
Formula:
C₆H₆N₂ Boiling Pt: 163…164 °C Density: 1.041 Flash Pt: 60 °C (140 °F) Storage Temperature: Freezer |
MDL Number:
MFCD00051591 CAS Number: 4177-16-6 EINECS: 224-045-2 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...