(2-Fluoro-4-methoxyphenyl)acetonitrile 97%, JRD
Supplier: Thermo Fisher Scientific
Danger
Synonyms: 2-(2-Fluoro-4-methoxyphenyl)acetonitrile
H30752.MD H30752.03
H30752.MDEA 67.1 EUR
H30752.MD H30752.03
(2-Fluoro-4-methoxyphenyl)acetonitrile 97%, JRD
(2-Fluoro-4-methoxyphenyl)acetonitrile
Purity: 97% CAS No.: 749934-29-0 Molecular Formula: C9H8FNO Formula Weight: 165.17 Refractive Index: 1.5131
Formula: C₉H₈FNO
MW: 165.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09038467
CAS Number: 749934-29-0
UN: 3276
ADR: 6.1 III