Acetonitrile with 0.1% (v/v) trifluoroacetic acid
Supplier: Roth Carl
Danger
CP02.1 CP02.2
ROTHCP02.1EA 119 EUR
ROTHCP02.1 ROTHCP02.2
Acetonitrile with 0.1% (v/v) trifluoroacetic acid
Acetonitrile with 0.1% (v/v) trifluoroacetic acid
Storage Temperature: Ambient
EINECS: 200-835-2
UN: 1648
ADR: 3,II