4-Dibenzofuranboronic acid

Supplier: Thermo Scientific

Synonyms: Dibenzofuran-4-boronic acid, Dibenzo[b,d]furan-4-ylboronic acid

AC35907.1 AC35907.10
MAYBAC35907.1EA 0 EUR
MAYBAC35907.1 MAYBAC35907.10
4-Dibenzofuranboronic acid
4-Dibenzofuranboronic acid
Formula: C₁₂H₉BO₃
MDL Number: MFCD00092336
CAS Number: 100124-06-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR