Thiazole-2-carbaldehyde 98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 2-Thiazolecarboxaldehyde, Thiazole-2-carboxaldehyde
377680010 377680050
ACRO377680010EA 68.5 EUR
ACRO377680010 ACRO377680050
Thiazole-2-carbaldehyde 98%
Thiazole-2-carbaldehyde
Formula: C₄H₃NOS
MW: 113.13 g/mol
Boiling Pt: 76…78 °C (24 mmHg)
Density: 1.288 g/cm³
Flash Pt: 68 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00142924
CAS Number: 10200-59-6
Order Now
Specification Test Results
Appearance Clear yellow to orange-red to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5730 to 1.5760 (20°C, 589 nm)
GC ≥97.5 %