4-Thiazolecarboxylic acid, Sigma-Aldrich®

Supplier: Merck

Synonyms: Thiazole-4-carboxylic acid

M-Clarity™ quality level = MQ100

633658-1G 633658-5G
SIAL633658-1GEA 27.2 EUR
SIAL633658-1G SIAL633658-5G
4-Thiazolecarboxylic acid, Sigma-Aldrich®
4-Thiazolecarboxylic acid
Formula: C₄H₃NO₂S
MDL Number: MFCD00623589
CAS Number: 3973-08-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR