Progesterone, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
P8783-1G P8783-25G P8783-5G
SIALP8783-1GEA 29.9 EUR
SIALP8783-1G SIALP8783-25G SIALP8783-5G
Progesterone, Sigma-Aldrich®
Progesterone
Formula: C₂₁H₃₀O₂
MDL Number: MFCD00003658
CAS Number: 57-83-0
EINECS: 200-350-6