(4-Acetamidophenyl)boronic acid 97+%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 4-Acetamidophenylboronic acid, 4-Acetamidobenzeneboronic acid, 4-Acetylaminophenylboronic acid
367610010 367610050
ACRO367610010EA 55.1 EUR
ACRO367610010 ACRO367610050
(4-Acetamidophenyl)boronic acid 97+%
(4-Acetamidophenyl)boronic acid
Formula: C₈H₁₀BNO₃
MW: 178.98 g/mol
Melting Pt: 215…220 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02179451
CAS Number: 101251-09-6
Order Now
Specification Test Results
Appearance Grey to tan Powder
Infrared spectrum Conforms
GC ≥97.0 %