Methyl-2-amino-4-chlorobenzoate

Supplier: Thermo Scientific

Synonyms: Methyl 2-amino-4-chlorobenzoate

BTB05014.1 BTB05014.10 BTB05014.50
MAYBBTB05014.1EA 0 EUR
MAYBBTB05014.1 MAYBBTB05014.10 MAYBBTB05014.50
Methyl-2-amino-4-chlorobenzoate
Methyl 2-amino-4-chlorobenzoate
Formula: C₈H₈ClNO₂
MDL Number: MFCD00017568
CAS Number: 5900-58-3
EINECS: 227-593-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR