6-Methoxy-2-naphthoic acid ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
L14610.03
L14610.06
L14610.03EA
52.6
EUR
L14610.03
L14610.06
6-Methoxy-2-naphthoic acid ≥98%
6-Methoxy-2-naphthoic acid
Purity: 98+%
CAS No.: 2471-70-7
Molecular Formula: C12H10O3
Formula Weight: 202.21
Melting Point: 192-194°
Beilstein Reference: 2211233
Formula:
C₁₂H₁₀O₃ Melting Pt: 192…194 °C Storage Temperature: Ambient |
MDL Number:
MFCD00092750 CAS Number: 2471-70-7 |