(S)-(+)-α-Cyclohexanemethylamine, Sigma-Aldrich®

Supplier: Merck

726796-100G 726796-25G
SIAL726796-100GEA 1120 EUR
SIAL726796-100G SIAL726796-25G
(S)-(+)-α-Cyclohexanemethylamine, Sigma-Aldrich®
(S)-(+)-α-Cyclohexanemethylamine
Formula: C₈H₁₇N
MDL Number: MFCD00066261
CAS Number: 17430-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR