General purpose gloves, ActivArmr® 47-200
Supplier: Ansell
47-200/10 47-200/7 47-200/8 47-200/9 47-200/6
ANSE47-200/10EA 53.6 EUR
ANSE47-200/10 ANSE47-200/7 ANSE47-200/8 ANSE47-200/9 ANSE47-200/6
General purpose gloves, ActivArmr® 47-200
Gloves
Cotton liner with teal nitrile coating for a better resistance to snags, punctures, abrasions and cuts when compared to ordinary cotton and string-knit gloves.
  • Ergonomically designed to be extremely comfortable, light and flexible
  • Superior dry grip, anti-static according to EN 1149
  • Lightweight nitrile coating repels oil and grease
  • Makes it easier and safer to handle smooth objects like boxes or appliances
  • Coating only on fingers and palm: Protects the working area, but leaves the back of the hand free to breathe
Ideal for assembling, inspecting and packing small or delicate parts, general maintenance, general material handling and shipping and receiving.
Certifications: EN 388:2016 (2111A), Cat. II