Hardened qualitative filter papers, standard grades, MN 1670, MN 1672 and MN 1674
Supplier: MACHEREY-NAGEL
470012 472007 472009 472012 474015 470005 470007 470009 470011 470015 470018 470024 470032 472005 472011 472015 472018 472024 472032 474005 474007 474009 474011 474012 474018 474024 474032 570005 570007 570009 570011 570012 570015 570018 570024 570032 572005 572007 572009 572011 572012 572015 572018 572024 572032 574005 574007 574009 574011 574012 574015 574018 574024 574032
MANA470012EA 26.5 EUR
MANA470012 MANA472007 MANA472009 MANA472012 MANA474015 MANA470005 MANA470007 MANA470009 MANA470011 MANA470015 MANA470018 MANA470024 MANA470032 MANA472005 MANA472011 MANA472015 MANA472018 MANA472024 MANA472032 MANA474005 MANA474007 MANA474009 MANA474011 MANA474012 MANA474018 MANA474024 MANA474032 MANA570005 MANA570007 MANA570009 MANA570011 MANA570012 MANA570015 MANA570018 MANA570024 MANA570032 MANA572005 MANA572007 MANA572009 MANA572011 MANA572012 MANA572015 MANA572018 MANA572024 MANA572032 MANA574005 MANA574007 MANA574009 MANA574011 MANA574012 MANA574015 MANA574018 MANA574024 MANA574032
Hardened qualitative filter papers, standard grades, MN 1670, MN 1672 and MN 1674
Filters Filter Papers Qualitative Filter Paper
Wet-strengthened analytical filter papers are made of refined pulp and linters and feature a content of α-cellulose of more than 95%. The smooth surface of these papers allows fibre-free filtration.
  • High wet-strength
  • For strongly acidic and basic media
Due to their high mechanical strength in wet condition they are particularly suited for applications, where the residue is removed from the filter with a spatula or a jet of water.
Grade MN 1670 is the fastest quantitative hardened filter paper with highest flow rate. It is used for example in filtration of samples with coarse or gelatinous particles.
Grade MN 1672 is a general purpose, hardened filter paper with medium speed and medium retention. It is used where high wet strength is needed in routine filtrations.
Grade MN 1674 is an ashless, slow filter paper with very fine particle retention. Standard hardened paper for filtrations of fine precipitates like barium sulphate or metastannic acid.