Pycnometer
Supplier: GLASWARENFABRIK KARL HECHT
42571024 42570008 42570016 42570024 42570032 42570040 42571008 42571308
HECH42571024EA 53.1 EUR
HECH42571024 HECH42570008 HECH42570016 HECH42570024 HECH42570032 HECH42570040 HECH42571008 HECH42571308
Pycnometer
Specific Gravity Bottles
Pycnometer, made of borosilicate glass 3.3, Gay-Lussac type.
  • With ground-in stopper
  • Good chemical resistance
  • Good temperature resistance
Certifications: DIN ISO 3507