3-Decyn-1-ol ≥97%
Supplier: Thermo Fisher Scientific
|
Warning
|
B20837.06
B20837.14
B20837.06EA
69
EUR
B20837.06
B20837.14
3-Decyn-1-ol ≥97%
3-Decyn-1-ol
Purity: 97%
CAS No.: 51721-39-2
Molecular Formula: CH3(CH2)5C≡CCH2CH2OH
Formula Weight: 154.25
Flash Point: 106°(222°F)
Refractive Index: 1.4590
Boiling Point: 129-130°/20mm
Beilstein Reference: 1750909
MDL No.: MFCD00040917
Formula:
C₁₀H₁₈O MW: 154.25 g/mol Boiling Pt: 129…130 °C (20 mmHg) Density: 0.871 Flash Pt: 106 °C (222 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040917 CAS Number: 51721-39-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...