2-Oxovaleric acid, Sigma-Aldrich®

Supplier: Merck

75950-25ML 75950-5ML
SIAL75950-25MLEA 571 EUR
SIAL75950-25ML SIAL75950-5ML
2-Oxovaleric acid, Sigma-Aldrich®
2-Oxovaleric acid
Formula: C₅H₈O₃
MDL Number: MFCD00066435
CAS Number: 1821-02-9
EINECS: 217-340-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR