1-Ethyl-1-methylpyrrolidinium bromide, Sigma-Aldrich®
Supplier: Merck
Synonyms:
1-Ethyl-1-methylpyrrolidin-1-ium bromide
713171-50G
713171-5G
SIAL713171-50GEA
165
EUR
SIAL713171-50G
SIAL713171-5G
1-Ethyl-1-methylpyrrolidinium bromide, Sigma-Aldrich®
1-Ethyl-1-methylpyrrolidinium bromide
Formula:
C₇H₁₆BrN |
MDL Number:
MFCD03095384 CAS Number: 69227-51-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...