Dibenzothiophene-4-boronic acid 95%
Supplier: Thermo Scientific
Warning
Synonyms: 4-Dibenzothiopheneboronic acid
AC35908.250 AC35908.1 359080010 359080050
MAYBAC35908.250EA 0 EUR
MAYBAC35908.250 MAYBAC35908.1 ACRO359080010 ACRO359080050
Dibenzothiophene-4-boronic acid 95%
Dibenzothiophene-4-boronic acid
Formula: C₁₂H₉BO₂S
MW: 228.08 g/mol
Melting Pt: 285…290 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01318182
CAS Number: 108847-20-7
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥94.0 %