Ytterbium(III) chloride ≥35% (w/w) in aqueous solution , 99.9% (REO, rare earth oxide basis)
Supplier: Thermo Fisher Scientific
42816.AD 42816.AK 42816.K2
42816.ADEA 74.1 EUR
42816.AD 42816.AK 42816.K2
Ytterbium(III) chloride ≥35% (w/w) in aqueous solution , 99.9% (REO, rare earth oxide basis)
Ytterbium (III) chloride
Purity: 99.9% CAS No.: 10361-91-8 Molecular Formula: YbCl3 Formula Weight: 279.40(anhy) Physical Form: Liquid Density: 1.48 MDL No.: MFCD00011288
Formula: Cl₃Yb
MW: 279.4 g/mol
Melting Pt: 860
Density: 1.48
Storage Temperature: Ambient
MDL Number: MFCD00011288
CAS Number: 10361-91-8
EINECS: 233-800-5
Merck Index: 13,10014