Chloroacetaldehyde 45% in aqueous solution, Sigma-Aldrich®
Supplier: Merck
10907-1KG 10907-25KG
SIAL10907-1KGEA 73.2 EUR
SIAL10907-1KG SIAL10907-25KG
Chloroacetaldehyde 45% in aqueous solution, Sigma-Aldrich®
Chloroacetaldehyde
Formula: C₂H₃ClO
MDL Number: MFCD00006992
CAS Number: 107-20-0
EINECS: 203-472-8