Chloroacetaldehyde 50% in aqueous solution, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
317276-5ML 317276-1L 317276-250ML
SIAL317276-5MLEA 22.2 EUR
SIAL317276-5ML SIAL317276-1L SIAL317276-250ML
Chloroacetaldehyde 50% in aqueous solution, Sigma-Aldrich®
Chloroacetaldehyde
Formula: C₂H₃ClO
MDL Number: MFCD00006992
CAS Number: 107-20-0
EINECS: 203-472-8