2,5-Dihydroxyterephthalic acid, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
382132-25G 382132-5G
SIAL382132-25GEA 492 EUR
SIAL382132-25G SIAL382132-5G
2,5-Dihydroxyterephthalic acid, Sigma-Aldrich®
2,5-Dihydroxyterephthalic acid
Formula: C₈H₆O₆
MDL Number: MFCD00132933
CAS Number: 610-92-4
EINECS: 210-239-4