Chloro(1,5-cyclooctadiene)iridium(I) dimer, Sigma-Aldrich®
Supplier: Merck
Synonyms: [Ir(cod)Cl]2, Chloro-1,5-cyclooctadiene iridium(I) dimer
M-Clarity™ quality level = MQ200
683094-2G 683094-500MG
SIAL683094-2GEA 370 EUR
SIAL683094-2G SIAL683094-500MG
Chloro(1,5-cyclooctadiene)iridium(I) dimer, Sigma-Aldrich®
Chloro(1,5-cyclooctadiene)iridium (I) dimer
Formula: C₁₆H₂₄Cl₂Ir₂
MDL Number: MFCD00012414
CAS Number: 12112-67-3
EINECS: 235-170-7