Chloro(1,5-cyclooctadiene)rhodium(I) dimer, Sigma-Aldrich®
Supplier: Merck
Synonyms: (1,5-Cyclooctadiene)rhodium chloride dimer
M-Clarity™ quality level = MQ100
227951-500MG 227951-5G
SIAL227951-500MGEA 491 EUR
SIAL227951-500MG SIAL227951-5G
Chloro(1,5-cyclooctadiene)rhodium(I) dimer, Sigma-Aldrich®
Chloro(1,5-cyclooctadiene)rhodium (I) dimer
Formula: C₁₆H₂₄Cl₂Rh₂
MW: 493.08 g/mol
MDL Number: MFCD00012415
CAS Number: 12092-47-6
EINECS: 235-157-6