(S)-(-)-2-Methyl-CBS-oxazaborolidine, Sigma-Aldrich®
Supplier: Merck
Synonyms: (S)-5,5-Diphenyl-2-methyl-3,4-propano-1,3,2-oxazaborolidine, (S)-Me-CBS Catalyst, (S)-3,3-Diphenyl-1-methyltetrahydro-1H,3H-pyrrolo[1,2-c][1,3,2]oxazaborole, (S)-3,3-Diphenyl-1-methylpyrrolidino[1,2-c]-1,3,2-oxazaborole
M-Clarity™ quality level = MQ100
649309-10G 649309-1G
SIAL649309-10GEA 314 EUR
SIAL649309-10G SIAL649309-1G
(S)-(-)-2-Methyl-CBS-oxazaborolidine, Sigma-Aldrich®
(S)-(-)-2-Methyl-CBS-oxazaborolidine
Formula: C₁₈H₂₀BNO
MW: 277.17 g/mol
MDL Number: MFCD00078439
CAS Number: 112022-81-8