Urinalysis kits, KOVA®
Supplier: Kova International
87154E 87137E 87135E 87139E
720-2420EA 979 EUR
720-2420 720-2418 720-2417 720-2419
Urinalysis kits, KOVA®
Clinical Diagnostic Supplies Clinical Reagents and Kits
Complete urinalysis kit for up to 1000 determinations.
  • 12 ml graduated non-sterile disposable centrifuge tubes manufactured of high-quality, break-resistant clear plastic and marked in 1 ml increments
  • Disposable plastic transfer pipettes that feature a unique lock for one-step, contamination-free decanting, retaining 1 ml of urine after centrifugation
  • Precision manufactured, 10-chambered disposable microscope slides made of optically clear plastic, the Glasstic® slide grid is in the focal plane of the specimen, allowing for use over a range of magnifications
Packaging: Contains 100 slides (10 chambered), 1000 KOVA Petters (pipettes with bulge) and 1000 KOVA tubes.