Membrane filters, PORAFIL® MV
Supplier: MACHEREY-NAGEL
65000020025 65000020047 65000020050 65000020090 65000020100 65000020142 65000020220 65000045013 65000045025 65000045047 65000045050 65000045090 65000045100 65000045142 65000045220 65000080013 65000080025 65000080047 65000080050 65000080090 65000080100 65000080142 65000080220 65000020013
MANA65000020025EA 54.8 EUR
MANA65000020025 MANA65000020047 MANA65000020050 MANA65000020090 MANA65000020100 MANA65000020142 MANA65000020220 MANA65000045013 MANA65000045025 MANA65000045047 MANA65000045050 MANA65000045090 MANA65000045100 MANA65000045142 MANA65000045220 MANA65000080013 MANA65000080025 MANA65000080047 MANA65000080050 MANA65000080090 MANA65000080100 MANA65000080142 MANA65000080220 MANA65000020013
Membrane filters, PORAFIL® MV
Filters Membrane Filters
Cellulose mixed ester membranes are reinforced with a polyester fabric. Ideal for gravimetric analysis and particularly suited for aqueous solutions.
  • Hydrophilic
  • Thermally stable at 125 °C
  • Suitable for contamination tests
These membranes are mechanically much more stable than PORAFIL® CM.