Membrane filters, PORAFIL® NC
Supplier: MACHEREY-NAGEL
6570020013 6570020025 6570020047 6570020050 6570020090 6570020100 6570020142 6570020220 6570045013 6570045025 6570045047 6570045050 6570045090 6570045100 6570045142 6570045220
MANA6570020013EA 48 EUR
MANA6570020013 MANA6570020025 MANA6570020047 MANA6570020050 MANA6570020090 MANA6570020100 MANA6570020142 MANA6570020220 MANA6570045013 MANA6570045025 MANA6570045047 MANA6570045050 MANA6570045090 MANA6570045100 MANA6570045142 MANA6570045220
Membrane filters, PORAFIL® NC
Filters Membrane Filters
PORAFIL® NC membranes are made from cellulose nitrate. They are easily wettable and suited for filtration of aqueous solutions.
  • Thermally stable to 125 °C