Membrane filters, PORAFIL® RC
Supplier: MACHEREY-NAGEL
659020013 659020025 659020047 659020050 659020100 659020142 659045013 659045025 659045047 659045050 659045100 659045142
MANA659020013EA 156 EUR
MANA659020013 MANA659020025 MANA659020047 MANA659020050 MANA659020100 MANA659020142 MANA659045013 MANA659045025 MANA659045047 MANA659045050 MANA659045100 MANA659045142
Membrane filters, PORAFIL® RC
Filters Membrane Filters
Regenerated cellulose membranes are used for filtration of solvent mixtures and ultrapurification and degassing of HPLC eluents.
  • Resistant to most organic solvents