Incubators, IN/INplus (natural convection)
Supplier: MEMMERT
IN110
IN110PLUS
IN160
IN160PLUS
IN260
IN260PLUS
IN30
IN30PLUS
IN450
IN450PLUS
IN55
IN55PLUS
IN75
IN750
IN750PLUS
IN75PLUS
IN160M
IN160MPLUS
IN450MPLUS
IN55M
IN750M
IN750MPLUS
IN260M
IN260MPLUS
IN55MPLUS
IN110M
IN30M
IN30MPLUS
IN75M
IN75MPLUS
IN110MPLUS
IN450M
390-0644EA
2051
EUR
390-0644
390-0645
390-0646
390-0647
390-0648
390-0649
390-0650
390-0651
390-0652
390-0653
390-0654
390-0655
390-0656
390-0657
390-0658
390-0659
MEMMIN160M
MEMMIN160MPLUS
MEMMIN450MPLUS
MEMMIN55M
MEMMIN750M
MEMMIN750MPLUS
MEMMIN260M
MEMMIN260MPLUS
MEMMIN55MPLUS
MEMMIN110M
MEMMIN30M
MEMMIN30MPLUS
MEMMIN75M
MEMMIN75MPLUS
MEMMIN110MPLUS
MEMMIN450M
Incubators, IN/INplus (natural convection)
Incubators
Standard Incubators
SPECIFICATIONS
Model | IN110 | IN160 | IN260 | IN30 | IN450 | IN55 | IN75 | IN750 | IN160 | IN55 | IN750 | IN260 | IN110 | IN30 | IN75 | IN450 |
Environmentally Preferable | ||||||||||||||||
Temperature fluctuation (time) | ≤±0,3 °C @ 37 °C | ≤±0,3 °C @ 37 °C | ≤±0,3 °C @ 37 °C | ≤±0,3 °C @ 37 °C | ≤±0,3 °C @ 37 °C | ≤±0,3 °C @ 37 °C | ≤±0,3 °C @ 37 °C | ≤±0,3 °C @ 37 °C | ≤±0,3 °C @ 37 °C | ≤±0,3 °C @ 37 °C | ≤±0,3 °C @ 37 °C | ≤±0,3 °C @ 37 °C | ≤±0,3 °C @ 37 °C | ≤±0,3 °C @ 37 °C | ≤±0,3 °C @ 37 °C | ≤±0,3 °C @ 37 °C |
Temperature variation (spatial) | ≤±0,6 °C @ 37 °C | ≤±0,6 °C @ 37 °C | ≤±0,6 °C @ 37 °C | ≤±0,6 °C @ 37 °C | ≤±0,6 °C @ 37 °C | ≤±0,6 °C @ 37 °C | ≤±0,6 °C @ 37 °C | ≤±0,6 °C @ 37 °C | ≤±0,6 °C @ 37 °C | ≤±0,6 °C @ 37 °C | ≤±0,6 °C @ 37 °C | ≤±0,6 °C @ 37 °C | ≤±0,6 °C @ 37 °C | ≤±0,6 °C @ 37 °C | ≤±0,6 °C @ 37 °C | ≤±0,6 °C @ 37 °C |
Capacity | 108 L | 161 L | 256 L | 32 L | 449 L | 53 L | 74 L | 749 L | 161 L | 53 L | 749 L | 256 L | 108 L | 32 L | 74 L | 449 L |
Convection type | Natural convection | Natural convection | Natural convection | Natural convection | Natural convection | Natural convection | Natural convection | Natural convection | ||||||||
Max. load per shelf | 20 kg | 20 kg | 20 kg | 20 kg | 30 kg | 20 kg | 20 kg | 30 kg | 20 kg | 20 kg | 30 kg | 20 kg | 20 kg | 20 kg | 20 kg | 30 kg |
Power consumption | 1400 W | 1600 W | 1700 W | 800 W | 1800 W | 1000 W | 1250 W | 2000 W | 1600 W | 1000 W | 2000 W | 1700 W | 1400 W | 800 W | 1250 W | 1800 W |
Temperature range | RT* +5…80 °C | RT* +5…80 °C | RT* +5…80 °C | RT* +5…80 °C | RT* +5…80 °C | RT* +5…80 °C | RT* +5…80 °C | RT* +5…80 °C | RT* +5…80 °C | RT* +5…80 °C | RT* +5…80 °C | RT* +5…80 °C | RT* +5…80 °C | RT* +5…80 °C | RT* +5…80 °C | RT* +5…80 °C |
Weight | 72 kg | 80 kg | 96 kg | 44 kg | 160 kg | 55 kg | 64 kg | 192 kg | 80 kg | 55 kg | 192 kg | 96 kg | 72 kg | 44 kg | 64 kg | 160 kg |
Services: Memmert IN/INplus incubators have a 3 year warranty period as standard. This is an additional 1 year extension to that 3 year period, giving 4 years in total.