Universal bare fused-silica capillaries
Supplier: Agilent
160-2644-5 160-2650-5 160-2660-5 190-0131 190-0231 190-0331 190-0431
HEWL160-2644-5EA 150 EUR
HEWL160-2644-5 HEWL160-2650-5 HEWL160-2660-5 HEWL190-0131 HEWL190-0231 HEWL190-0331 HEWL190-0431
Universal bare fused-silica capillaries
Electrophoresis Systems Capillary Electrophoresis Systems
Universal Bare Fused-Silica Capillaries have a window, 750 mm effective length and 363 μm Ext.Ø, fitting into any CE instrument.