2-Bromoisobutyryl bromide 98%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
2-Bromo-2-methylpropionyl bromide
403090050
403091000
403095000
ACRO403090050EA
27.2
EUR
ACRO403090050
ACRO403091000
ACRO403095000
2-Bromoisobutyryl bromide 98%
2-Bromoisobutyryl bromide
Formula:
C₄H₆Br₂O Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000122 CAS Number: 20769-85-1 EINECS: 244-017-3 UN: 3265 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to light brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5070 to 1.5100 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...