2,6-Diacetylpyridine ≥99%
Supplier: Thermo Fisher Scientific
Warning
B25678.03 B25678.06 B25678.14
B25678.03EA 53.4 EUR
B25678.03 B25678.06 B25678.14
2,6-Diacetylpyridine ≥99%
2,6-Diacetylpyridine
Purity: 99% CAS No.: 1129-30-2 Molecular Formula: C9H9NO2 Formula Weight: 163.18 Melting Point: 78-82° MDL No.: MFCD00006304
Formula: C₉H₉NO₂
Melting Pt: 78…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00006304
CAS Number: 1129-30-2
EINECS: 214-442-9