Lithiumbis(trimethylsilyl)amide 1 M in THF/ethylbenzene, AcroSeal®
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Lithiumbis(triméthylsilyl)amide, HMDS-Li, N-Lithiohexamethyldisilazane, HMDS lithium salt
347701000 347708000
347701000.EA 61.2 EUR
347701000. 347708000.
Lithiumbis(trimethylsilyl)amide 1 M in THF/ethylbenzene, AcroSeal®
Lithium bis(trimethylsilyl)amide
Formula: C₆H₁₈LiNSi₂
MW: 167.33 g/mol
Density: 0.89 g/cm³
Flash Pt: –21 °C
Storage Temperature: Ambient
MDL Number: MFCD00008261
CAS Number: 4039-32-1
EINECS: 223-725-6
UN: 2924
ADR: 3 II
Order Now
Specification Test Results
Appearance Yellow to brown Solution
Titration Active base 18 to 22 % (as LiNSi)