Methyl dimethoxyacetate ≥97%
Supplier: Thermo Fisher Scientific
Warning
|
A12171.09
A12171.18
A12171.30
A12171.09EA
71.6
EUR
A12171.09
A12171.18
A12171.30
Methyl dimethoxyacetate ≥97%
Methyl dimethoxyacetate
Purity: 97+%
CAS No.: 89-91-8
Molecular Formula: (CH3O)2CHCO2CH3
Formula Weight: 134.13
Flash Point: 63°(145°F)
Refractive Index: 1.4050
Boiling Point: 160-163°
Beilstein Reference: 1757582
MDL No.: MFCD00008484
Formula:
C₅H₁₀O₄ Boiling Pt: 160…163 °C Density: 1.098 Flash Pt: 63 °C (145 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008484 CAS Number: 89-91-8 EINECS: 201-950-0 |